Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dyphylline, USP, 98-102%, Spectrum™

Click to view available options
Quantity:
25 g
100 g
500 g
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
479-18-5
C10H14N4O4
254.25
KSCFJBIXMNOVSH-UHFFFAOYNA-N
7-(2,3-dihydroxypropyl)-1,3-dimethyl-2,3,6,7-tetrahydro-1H-purine-2,6-dione
CN1C2=C(N(CC(O)CO)C=N2)C(=O)N(C)C1=O
479-18-5
C10H14N4O4
254.25
KSCFJBIXMNOVSH-UHFFFAOYNA-N
7-(2,3-dihydroxypropyl)-1,3-dimethyl-2,3,6,7-tetrahydro-1H-purine-2,6-dione
CN1C2=C(N(CC(O)CO)C=N2)C(=O)N(C)C1=O

D2202, 479-18-5, C10H14N4O4

Spectrum™ Dyphylline, USP belongs to a class of drugs known as xanthines that are used to treat certain respiratory disorders. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Amber Glass Bottle
Ignition Residue 0.0015
Melting Range 160-164°C
Quantity 500 g
CAS Max % 1
Percent Purity 98 to 102%