Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DYRK1A Recombinant Human Protein Non-distribution product as customer accommodation.

DYRK1A Recombinant Human Protein

$433.00

Specifications

Accession Number NP_001387
Common Name DYRK1A Recombinant Human Protein
Concentration 0.16mg/mL
Expression System Baculovirus
Gene Alias DYRK1A, DYRK1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PV3785 Non-distribution product as customer accommodation. View Documents LIFE TECHNOLOGIES
PV3785
10μg Each for $433.00
Add to cart
 
Description

Description

DYRK1A is a dual specificity kinase involved in cell proliferation. The gene for DYRK1A is located in the Down syndrome critical region of chromosome 21, and has been implicated in the pathogenesis of this disease.
Specifications

Specifications

NP_001387
0.16mg/mL
DYRK1A, DYRK1
DYRK1A Recombinant Human Protein
DYRK1A Recombinant Human Protein
For research use only. Not for use in diagnostic procedures
–80°C
Human
DYRK1A Recombinant Human Protein
Baculovirus
113kDa
1 Each
85% by SDS-PAGE
2880nmoles of phosphate transferred to DYRKtide peptide substrate (RRRFRPASPLRGPPK) per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 0.83μg/mL
Kinase assay
GST Tag

For Research Use Only. Not for use in diagnostic procedures.

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit