Learn More
DYRK2 Mouse anti-Human, Clone: OTI1G8, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF810286
Catalog No. 50-166-9989
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
DYRK2, or dual specificity tyrosine-phosphorylation-regulated kinase 2, has a role in the regulation of cellular growth and/or development. In response to DNA damage, DYRK2 regulates p53 phosphorylation on Ser-46 to induce apoptosis. DYRK2 has been proposed to inactivate GYS1 by phosphorylation at Ser-641, and potentially a second phosphorylation site, thus regulating glycogen synthesis. DYRK2 also phosphorylates EIF2B5 at Ser-544, enabling its subsequent phosphorylation and inhibition by GSK3, and may play a more general role in the priming of GSK3 substrates.
Specifications
Specifications
DYRK2
Monoclonal
Unconjugated
DYRK2
Lyophilized
DYRK2
Human recombit protein fragment corresponding to amino acids 1-210 of human DYRK2 produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Western Blot
OTI1G8
PBS with 8% trehalose and no preservative; pH 7.3
Q92630
dual specificity tyrosine phosphorylation regulated kinase 2
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
8445