Learn More
Dysbindin1 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A303360A

Description
The recommended shelf life for this product is 1 year from date of receipt.
This gene encodes a protein that may play a role in organelle biogenesis associated with melanosomes, platelet dense granules, and lysosomes. A similar protein in mouse is a component of a protein complex termed biogenesis of lysosome-related organelles complex 1 (BLOC-1), and binds to alpha- and beta-dystrobrevins, which are components of the dystrophin-associated protein complex (DPC). Mutations in this gene are associated with Hermansky-Pudlak syndrome type 7. This gene may also be associated with schizophrenia. Multiple transcript variants encoding distinct isoforms have been identified for this gene.Specifications
Dysbindin1 | |
Polyclonal | |
Unconjugated | |
DTNBP1 | |
Dysbindin; Dystrobrevin-binding protein 1; Hermansky-Pudlak syndrome 7 protein homolog; Hps7-like protein; Dtnbp1; Sdy | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
84062 | |
4° C | |
Liquid |
Immunoprecipitation, Western Blot | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q96EV8 | |
DTNBP1 | |
Between 301 and 351 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.