Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dysferlin Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549738

Catalog No. PA549738


Add to cart

Description

Description

The antibody detects endogenous levels of total Dysferlin protein.

Dysferlin is the protein product of the 2p13 gene that is defective in patients with Limb-Girdle Muscular Dystrophy type 2B (LGMD2B) and Miyoshi Myopathy (MM). Dysferlin is normally localized to the muscle plasma membrane. In patients with LGMD2B and MM, immunoreactivity to dysferlin is severely reduced or lost, depending on the type of mutation. This antibody is used for the characterization of LGMD2B and MM.
Specifications

Specifications

Dysferlin
Polyclonal
Unconjugated
DYSF
Liquid
DYSF
Synthesized peptide derived from human Dysferlin.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
O75923, Q9ESD7
FER1L1, FLJ00175, FLJ90168, LGMD2B
Rabbit
IgG
100 μL
-20°C
Primary
26903, 8291
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit