Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPP1R27 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548520

Catalog No. PA548520

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Inhibits phosphatase activity of protein phosphatase 1 (PP1) complexes.
Specifications

Specifications

PPP1R27
Polyclonal
PBS with 0.09% sodium azide
Q86WC6
Protein phosphatase 1 regulatory subunit 27, Dysferlin-interacting protein 1, Toonin, PPP1R27, DYSFIP1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
116729
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
PPP1R27
Liquid
PPP1R27
KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human DYSFIP1.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit