Alfa Aesar™ Dysprosium(III) acetate tetrahydrate, REacton™, 99.9% (REO)

Manufacturer: Alfa Aesar™ 1459514

Quantity 25g

 View more versions of this product

Catalog No. AA1459514

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 15280-55-4
    Formula Weight 411.68 (339.62 Anhydrous)
    MDL Number MFCD00013033
    Chemical Name or Material Dysprosium(III) acetate tetrahydrate
    Grade Reacton™

    Specifications

    Formula Weight 411.68 (339.62 Anhydrous)
    MDL Number MFCD00013033
    Physical Form Crystalline Aggregates
    CAS 15280-55-4
    TSCA TSCA
    Odor Odorless
    Melting Point 120°C (dec.)
    Quantity 25g
    Chemical Name or Material Dysprosium(III) acetate tetrahydrate
    Assay 99.9%, (REO)
    Grade Reacton™

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit