Alfa Aesar™ Dysprosium(III) bromide, ultra dry, 99.99% (REO)

Manufacturer: Alfa Aesar™ 3861106

Quantity 5g

 View more versions of this product

Catalog No. AA3861106

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 14456-48-5
    Formula Weight 402.23
    MDL Number MFCD00016069
    Chemical Name or Material Dysprosium(III) bromide, Ultra dry

    Specifications

    Formula Weight 402.23
    Packaging Ampule
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Quantity 5g
    Sensitivity Hygroscopic
    MDL Number MFCD00016069
    Health Hazard 1 Warning
    Physical Form Powder
    CAS 14456-48-5
    Boiling Point 1480°C
    Melting Point 881°C
    Solubility Information Soluble in water
    TSCA TSCA
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Storage Note 1
    • Ampouled under argon
    Chemical Name or Material Dysprosium(III) bromide, Ultra dry
    Assay 99.99%, (REO)

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    • Ampouled under argon

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit