Alfa Aesar™ Dysprosium(III) carbonate tetrahydrate, 99.9% (REO)

Manufacturer: Alfa Aesar™ 1528618

Quantity 50g

 View more versions of this product

Catalog No. AA1528618

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 38245-35-1
    Formula Weight 577.06 (505.03 Anhydrous)
    MDL Number MFCD00150229
    Chemical Name or Material Dysprosium(III) carbonate tetrahydrate

    Specifications

    Formula Weight 577.06 (505.03 Anhydrous)
    Physical Form Powder
    Sensitivity Hygroscopic
    MDL Number MFCD00150229
    Quantity 50g
    TSCA TSCA
    CAS 38245-35-1
    Chemical Name or Material Dysprosium(III) carbonate tetrahydrate
    Assay 99.9%, (REO)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit