Alfa Aesar™ Dysprosium(III) chloride, ultra dry, 99.99% (REO)

Manufacturer: Alfa Aesar™ 4711414

Quantity 25g

 View more versions of this product

Catalog No. AA4711414

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 10025-74-8
    Formula Weight 268.86
    MDL Number MFCD00010984
    Merck Index 14,3482
    Chemical Name or Material Dysprosium(III) chloride, Ultra dry

    Specifications

    Formula Weight 268.86
    Packaging Ampule
    Health Hazard 3 GHS P Statement
    P280-P264-P305+P351+P338-P337+P313
    Wear protective gloves/protective clothing/eye protection/face protection.
    Wash thoroughly after handling.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    If eye irritation persists: Get medical advice/attention.
    CAS 10025-74-8
    TSCA TSCA
    Physical Form Beads
    MDL Number MFCD00010984
    Merck Index 14,3482
    Density 3.67g/mL
    Mesh Size −10 Mesh
    Sensitivity Hygroscopic
    Quantity 25g
    Storage Note 1
    • Ampouled under argon
    Odor Odorless
    Melting Point 718°C
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H319
    Causes serious eye irritation.
    Chemical Name or Material Dysprosium(III) chloride, Ultra dry
    Assay 99.99%, (REO)

    GHS H Statement
    H319
    Causes serious eye irritation.

    GHS P Statement
    P280-P264-P305+P351+P338-P337+P313
    Wear protective gloves/protective clothing/eye protection/face protection.
    Wash thoroughly after handling.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    If eye irritation persists: Get medical advice/attention.

    • Ampouled under argon

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit