Alfa Aesar™ Dysprosium(III) fluoride, anhydrous, REacton™, 99.9% (REO)

Manufacturer: Alfa Aesar™ 1365614

Quantity 25g

 View more versions of this product

Catalog No. AA1365614

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13569-80-7
    Formula Weight 219.5
    MDL Number MFCD00016071
    Chemical Name or Material Dysprosium(III) fluoride, Anhydrous
    Grade Reacton™

    Specifications

    Formula Weight 219.5
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Odor Odorless
    CAS 13569-80-7
    Melting Point 1360°C
    Quantity 25g
    Physical Form Crystalline Lump
    Sensitivity Hygroscopic
    MDL Number MFCD00016071
    Boiling Point 2200°C
    Density 7.5g/mL
    TSCA TSCA
    Color White
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Chemical Name or Material Dysprosium(III) fluoride, Anhydrous
    Assay 99.9%, (REO)
    Grade Reacton™

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit