Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Dysprosium(III) isopropoxide, 99.9% (REO), 5% w/v in toluene/isopropanol

Manufacturer:  Alfa Aesar™ 36585AA

Quantity 5mL

 View more versions of this product

Catalog No. AA36585AA


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

6742-68-3
339.76
UN1993
Dy[OCH(CH3)2]3
MFCD00049784
Dysprosium(III) isopropoxide

Specifications

5% w/v in toluene/isopropanol
5mL
Liquid
339.76
UN1993
Dysprosium(III) isopropoxide
Moisture Sensitive
6742-68-3
Dy[OCH(CH3)2]3
MFCD00049784
Brown
99.9%, (REO)

Safety and Handling

SDS & Safety Documents

GHS H Statement
H225-H304-H361-H373-H315-H319-H336
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
Causes serious eye irritation.
May cause drowsiness or dizziness.

GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit