Alfa Aesar™ Dysprosium(III) oxalate decahydrate, REacton™, 99.9% (REO)

Manufacturer: Alfa Aesar™ 1459322

Quantity 100g

 View more versions of this product

Catalog No. AA1459322

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 24670-07-3
    Linear Formula Dy2(C2O4)3·10H2O
    Formula Weight 769.21 (589.06 Anhydrous)
    MDL Number MFCD00150482
    UN Number UN3288
    Chemical Name or Material Dysprosium(III) oxalate decahydrate
    Grade Reacton™

    Specifications

    Formula Weight 769.21 (589.06 Anhydrous)
    Melting Point 40°C (in -H2O)
    Health Hazard 3 GHS P Statement
    P280-P301+P312-P312-P363-P322-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Wash contaminated clothing before reuse.
    Specific measures (see label).
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Color White
    Physical Form Powder
    Sensitivity Hygroscopic
    Health Hazard 1 Warning
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
    TSCA TSCA
    Linear Formula Dy2(C2O4)3·10H2O
    MDL Number MFCD00150482
    CAS 24670-07-3
    UN Number UN3288
    Quantity 100g
    Health Hazard 2 GHS H Statement
    H302-H312
    Harmful if swallowed.
    Harmful in contact with skin.
    Chemical Name or Material Dysprosium(III) oxalate decahydrate
    Assay 99.9%, (REO)
    Grade Reacton™

    GHS H Statement
    H302-H312
    Harmful if swallowed.
    Harmful in contact with skin.

    GHS P Statement
    P280-P301+P312-P312-P363-P322-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Wash contaminated clothing before reuse.
    Specific measures (see label).
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit