Learn More
Dysprosium(III) oxide, REacton™, 99.9% (REO), Thermo Scientific Chemicals
Description
In cermets used as nuclear reaction control rodsDysprosium trioxide is used with nickel in cermets, which are used as nuclear reactor control rods. It is also used in ceramics, glass, phosphors, lasers and dysprosium metal halide lamps. It is an important raw material to prepare dysprosium(III) salts by reacting with acids.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 2340°C |
Color | White |
Physical Form | Powder |
Quantity | 50 g |
Assay Percent Range | 99.9% (REO) |
Solubility Information | Insoluble in water. |
Formula Weight | 373 |
Sensitivity | Hygroscopic |
Appearance | White powder or lump |
Density | 7.81 g/cm3 |
Show More |
Safety and Handling
EINECSNumber | 215-164-0 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.