Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Dysprosium(III) oxide, nanoneedles, 99.5% (REO)

Manufacturer:  Alfa Aesar™ 4466409

Quantity 10g

 View more versions of this product

Catalog No. AA4466409

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

1308-87-8
373
14,3482
Dy2O3
MFCD00010986
Dysprosium (III) oxide

Specifications

1308-87-8
Dy2O3
MFCD00010986
Soluble in acids and alcohol
20 to 30nm x 200 to 300nm
2340°C
Dysprosium (III) oxide
Nanoneedles
373
14,3482
7.81g/mL
18 to 22m2/g
10g
99.50%, (REO)

Safety and Handling

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit