Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dysprosium powder, -200 mesh, 99.9% (REO), Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under Argon
Specifications
Color | Silver |
Quantity | 25 g |
UN Number | UN3089 |
Percent Purity | 99.9% |
Mesh Size | −200 Mesh |
Odor | Odorless |
Sensitivity | Air and moisture sensitive |
Form | Powder |
Assay | (REO) |
Chemical Name or Material | Dysprosium powder |
Safety and Handling
Hazard Category | H228-H260 |
Hazard Statement | GHS H Statement H228-H260 Flammable solid. In contact with water releases flammable gases which may ignite spontaneously. |
Precautionary Statement | P210-P223-P231+P232-P240-P241-P280-P335+P334-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: METAL POWDERS, FLAMMABLE, N.O.S. |
EINECSNumber | 231-073-9 |
TSCA | Yes |
storageNote1 |
|
Recommended Storage | Ambient temperatures; Store under Argon |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction