Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dysprosium Rod, 6.35mm (0.25 in.) dia, 99.7% (Metals basis excluding Ta), Alfa Aesar™
Click to view available options
Quantity:
10mm
25mm
50mm
100mm
Specifications
Assay Percent Range | 99.7% (Metals basis excluding Ta) |
Appearance | Gray to silver rod |
Physical Form | ≈0.27g/mm,Cast Rod,diameter is approximate |
Odor | Odorless |
Color | Gray to Silver |
Quantity | 100mm |
Chemical Name or Material | Dysprosium |
Safety and Handling
EINECSNumber : 231-073-9
TSCA : Yes
Recommended Storage : Ambient temperatures