Alfa Aesar™ Dysprosium sputtering target, 50.8mm dia x 1.59mm thick

Manufacturer: Alfa Aesar™ 45619KS

Quantity 1 ea.

 View more versions of this product

Catalog No. AA45619KS

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7429-91-6
    MDL Number MFCD00010982
    Chemical Name or Material Dysprosium sputtering target, 50.8mm dia. x 1.59mm thick

    Specifications

    Quantity 1 ea.
    MDL Number MFCD00010982
    Odor Odorless
    TSCA TSCA
    CAS 7429-91-6
    Chemical Name or Material Dysprosium sputtering target, 50.8mm dia. x 1.59mm thick

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit