Learn More
Dystrobrevin alpha Antibody (OTI1B2), DyLight 405, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP270591V
Catalog No. NBP270591V
Add to Cart
Description
Description
Dystrobrevin alpha Monoclonal specifically detects Dystrobrevin alpha in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
Dystrobrevin alpha
Monoclonal
DyLight 405
50mM Sodium Borate with 0.05% Sodium Azide
DTNA
Full length recombinant protein of human Dystrobrevin alpha (NP_116761) produced in HEK293T cells.
0.1 mL
1837.0
Store at 4°C in the dark.
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
OTI1B2
Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence
Alpha-dystrobrevin, D18S892EDTN, DRP3FLJ96209, DTN-1, DTN-2, DTN-3, DTN-A, dystrobrevin alpha, dystrobrevin, alpha, Dystrophin-related protein 3, LVNC1
Mouse
Affinity Purified
Primary
Human, Mouse, Rat
IgG2a
Product Suggestions
Product Suggestions