Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dystrobrevin alpha Antibody (OTI1B2), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270591MFV610
Description
Dystrobrevin alpha Monoclonal antibody specifically detects Dystrobrevin alpha in Human, Mouse, Rat samples. It is validated for Flow CytometrySpecifications
Dystrobrevin alpha | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Mouse | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Flow Cytometry | |
OTI1B2 | |
Flow Cytometry | |
Alpha-dystrobrevin, D18S892EDTN, DRP3FLJ96209, DTN-1, DTN-2, DTN-3, DTN-A, dystrobrevin alpha, dystrobrevin, alpha, Dystrophin-related protein 3, LVNC1 | |
Full length human recombinant protein of human DTNA (NP_116761) produced in HEK293T cell. | |
0.1 mL | |
Cell Biology, Cytoskeleton Markers, Neuroscience, Signal Transduction | |
1837 | |
Store at 4°C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction