Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dystrobrevin alpha Mouse anti-Human, Janelia Fluor™ 646, Clone: OTI1B2, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270591JF646

Catalog No. N270591J646


Add to cart

Description

Description

Dystrobrevin alpha Monoclonal antibody specifically detects Dystrobrevin alpha in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunofluorescence.
Specifications

Specifications

Dystrobrevin alpha
OTI1B2
Flow Cytometry, Immunocytochemistry/Immunofluorescence
Affinity Purified, Conjugated
DTNA
Full length recombinant protein of human Dystrobrevin alpha (NP_116761) produced in HEK293T cells.
Immunogen affinity purified
Store at 4°C in the dark.
Monoclonal
Human
Flow Cytometry, Immunocytochemistry, Immunofluorescence
Janelia Fluor 646
50mM Sodium Borate with 0.05% Sodium Azide
Alpha-dystrobrevin, D18S892EDTN, DRP3FLJ96209, DTN-1, DTN-2, DTN-3, DTN-A, dystrobrevin alpha, dystrobrevin, alpha, Dystrophin-related protein 3, LVNC1
Mouse
IgG2a
0.1 mL
Primary
1837.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit