Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dystrobrevin alpha Antibody (OTI1B2), PE/Cy5.5, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270591PECY55
Description
Dystrobrevin alpha Monoclonal specifically detects Dystrobrevin alpha in Human, Mouse, Rat samples. It is validated for Flow Cytometry.Specifications
Dystrobrevin alpha | |
Monoclonal | |
PE-Cyanine5.5 | |
PBS with 0.05% Sodium Azide | |
DTNA | |
Full length recombinant protein of human Dystrobrevin alpha (NP_116761) produced in HEK293T cells. | |
0.1 mL | |
1837.0 | |
Store at 4°C in the dark. Do not freeze. |
Flow Cytometry | |
OTI1B2 | |
Flow Cytometry | |
Alpha-dystrobrevin, D18S892EDTN, DRP3FLJ96209, DTN-1, DTN-2, DTN-3, DTN-A, dystrobrevin alpha, dystrobrevin, alpha, Dystrophin-related protein 3, LVNC1 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG2a |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction