Learn More
Dystrobrevin beta Antibody (OTI3A12), PE, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP270598PE
Catalog No. NBP270598PE
Add to Cart
Description
Description
Dystrobrevin beta Monoclonal specifically detects Dystrobrevin beta in Human, Mouse, Rat samples. It is validated for Flow Cytometry.
Specifications
Specifications
Dystrobrevin beta
Monoclonal
PE
PBS with 0.05% Sodium Azide
DTNB
Full length recombinant protein of human Dystrobrevin beta (NP_899205) produced in HEK293T cells.
0.1 mL
1838.0
Store at 4°C in the dark.
Flow Cytometry
OTI3A12
Flow Cytometry
beta-dystrobrevin, DTN-B, dystrobrevin beta, dystrobrevin, beta, MGC17163, MGC57126
Mouse
Affinity Purified
Primary
Human, Mouse, Rat
IgG1
Recommendations
Recommendations