Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dystrobrevin beta Mouse anti-Human, PE/Atto594, Clone: OTI3A12, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270598PEATT594

Catalog No. N270598P594


Add to cart

Description

Description

Dystrobrevin beta Monoclonal antibody specifically detects Dystrobrevin beta in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

Dystrobrevin beta
OTI3A12
Flow Cytometry
Affinity Purified, Conjugated
DTNB
Full length recombinant protein of human Dystrobrevin beta (NP_899205) produced in HEK293T cells.
Immunogen affinity purified
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human
Flow Cytometry
PE/ATTO 594
PBS with 0.05% Sodium Azide
beta-dystrobrevin, DTN-B, dystrobrevin beta, dystrobrevin, beta, MGC17163, MGC57126
Mouse
IgG1
0.1 mL
Primary
1838.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit