Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dystroglycan Rabbit anti-Human, DyLight 405, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP300293V

Catalog No. NB065449


Add to cart

Description

Description

Dystroglycan Polyclonal specifically detects Dystroglycan in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

Dystroglycan
DyLight 405
50mM Sodium Borate
A3a, AGRNR, DAG156DAG, dystroglycan, dystroglycan 1 (dystrophin-associated glycoprotein 1), Dystrophin-associated glycoprotein 1
Produced in rabbits immunized with purified, recombinant Dystroglycan (NP_004384.4; Met1-Val749).
Affinity purified
RUO
Store at 4°C in the dark.
Polyclonal
Human
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Purified
Rabbit
IgG
0.1 mL
Breast Cancer, Cancer, Phospho Specific
Primary
1605
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit