Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ Dystrophin Ab-1, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry, immunofluorescence, affinity purification and electron microscopy applications.

Specifications

Antigen Dystrophin Ab-1
Clone 1808
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
MS486R1
View Documents
Thermo Scientific™
MS486R1
Ready-to-use for IHC (UVLP) 1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
MS486R7
View Documents
Thermo Scientific™
MS486R7
Ready-to-use for IHC (UVLP) 7mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Ensure accurate, reproducible results in immunohistochemistry, immunofluorescence, affinity purification and electron microscopy applications with Thermo Scientific™ Dystrophin Ab-1, Mouse Monoclonal Antibody.

Dystrophin is a member of the spectrin/α-actinin family of actin-binding, triple helix rod-containing proteins. It is absent or greatly reduced in individuals with the X-linked recessive Duchenne's muscular dystrophy disorder, as well as in mice with the mdx (murine muscular dystrophy) mutation.

Host Species: Mouse

Clone: 1808

Isotype: IgG1

Species Reactivity: Human, Mouse, Rat, Chicken, Xenopus and Torpedo. Others not known.

Epitope: Not determined

Immunogen: Acetylcholine receptor (AChR)-enriched membranes and peripheral membrane proteins from Torpedo nobiliana electric organ

Molecular Weight: 427 kDa

Positive Control: Skeletal muscle

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Immunofluorescence
  • Affinity Purification
  • Electron Microscopy

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Dystrophin Ab-1
Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
RUO
Primary
IgG1
1808
Monoclonal
Murine
Muscle Biology
Acetylcholine receptor (AChR)-enriched membranes and peripheral membrane proteins from Torpedo nobiliana electric organ
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit