Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dystrophin Mouse anti-Human, Clone: DMD/3677, Novus Biologicals™
Mouse Monoclonal Antibody
$265.00
Specifications
Antigen | Dystrophin |
---|---|
Clone | DMD/3677 |
Concentration | 0.2 mg/ml |
Dilution | ELISA, Immunohistochemistry-Paraffin 1-2 ug/ml |
Applications | ELISA, Immunohistochemistry (Paraffin) |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB122784
|
Novus Biologicals
NBP30765520UG |
20 μg |
Each of 1 for $265.00
|
|
|||||
Description
Dystrophin Monoclonal specifically detects Dystrophin in Human samples. It is validated for ELISA, Immunohistochemistry-Paraffin.Specifications
Dystrophin | |
0.2 mg/ml | |
ELISA, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Cell Biology, Cellular Markers | |
10 mM PBS with 0.05% BSA | |
1756 | |
IgG κ | |
Protein A or G purified |
DMD/3677 | |
ELISA, Immunohistochemistry-Paraffin 1-2 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
Human | |
BMDDXS272, CMD3B, DXS142, DXS164, DXS164, DXS206, DXS230, DXS239, DXS268, DXS269, DXS270, DXS272, DXS206, DXS230, DXS239, DXS268, DXS269, DXS270, dystrophin, dystrophin (muscular dystrophy, Duchenne and Becker types), includes DXS142 | |
A recombinant fragment (around aa 114-263) of human Dystrophin protein (exact sequence is proprietary) | |
Primary | |
Store at 4°C. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title