Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dystrophin, Mouse anti-Human, DyLight 755, Clone: DMD/3241, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP279910IR
Description
Dystrophin Monoclonal antibody specifically detects Dystrophin in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin).Specifications
Dystrophin | |
DMD/3241 | |
ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
Conjugated, Purified | |
DMD | |
A recombinant fragment (around aa 114-263) of human Dystrophin protein (exact sequence is proprietary) | |
Protein A or G purified | |
RUO | |
Store at 4°C in the dark. | |
Monoclonal |
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
BMDDXS272, CMD3B, DXS142, DXS164, DXS164, DXS206, DXS230, DXS239, DXS268, DXS269, DXS270, DXS272, DXS206, DXS230, DXS239, DXS268, DXS269, DXS270, dystrophin, dystrophin (muscular dystrophy, Duchenne and Becker types), includes DXS142 | |
Mouse | |
IgG1 Kappa | |
0.1 mL | |
Cell Biology, Cellular Markers | |
Primary | |
Human |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title