Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dystrophin, Mouse anti-Human, Janelia Fluor 549, Clone: DMD/3241, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP279910JF549

Catalog No. NB012378


Add to cart

Description

Description

Dystrophin Monoclonal antibody specifically detects Dystrophin in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin).
Specifications

Specifications

Dystrophin
DMD/3241
Immunohistochemistry, Immunohistochemistry-Paraffin
Conjugated, Purified
DMD
A recombinant fragment (around aa 114-263) of human Dystrophin protein (exact sequence is proprietary)
Protein A or G purified
RUO
Store at 4°C in the dark.
Monoclonal
Immunohistochemistry, Immunohistochemistry (Paraffin)
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
BMDDXS272, CMD3B, DXS142, DXS164, DXS164, DXS206, DXS230, DXS239, DXS268, DXS269, DXS270, DXS272, DXS206, DXS230, DXS239, DXS268, DXS269, DXS270, dystrophin, dystrophin (muscular dystrophy, Duchenne and Becker types), includes DXS142
Mouse
IgG1 Kappa
0.1 mL
Cell Biology, Cellular Markers
Primary
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit