Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ Dystrophin, Rabbit Polyclonal Antibody

Obtain accurate, reproducible results in immunohistochemistry experiments with Thermo Scientific™ Dystrophin, Rabbit Polyclonal Antibody.

$629.50 - $879.00

Specifications

Antigen Dystrophin
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 5
Catalog Number Mfr. No. Format Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Format Quantity Price Quantity & Availability  
RB9024P
View Documents
Thermo Scientific™
RB9024P
Concentrated 1mL
Each for $879.00
Add to cart
 
RB9024P1
View Documents
Thermo Scientific™
RB9024P1
Concentrated 500μL
Each for $629.50
Add to cart
 
RB9024P0
View Documents
Thermo Scientific™
RB9024P0
Concentrated 100μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
RB9024R1
View Documents
Thermo Scientific™
RB9024R1
Prediluted 1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
RB9024R7
View Documents
Thermo Scientific™
RB9024R7
Prediluted 7mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description

Description

Obtain accurate, reproducible results in immunohistochemistry experiments with Thermo Scientific™ Dystrophin, Rabbit Polyclonal Antibody, one marker in a comprehensive antibody portfolio that addresses all major disease states, ensuring the ideal tests are always available.

Dystrophin is the 427kDa protein product of the DMB/BMD gene located on the X chromosome at position Xp21. Western blotting and immuohistochemistry are the two established methods for the detection of abnormalities of dystrophin expression in muscle biopsies. Dystrophin abnormalities are thought to occur in 100% of patients with DMD/BMD, although genetic abnormalities may only be detected in up to 65% of cases.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human and Rat. Others not tested.

Epitope: C-terminal

Immunogen: A synthetic peptide corresponding to C-terminal of human Dystrophin

Molecular Weight: 427 kDa

Positive Control: Skeletal muscle

Cellular Localization: Cell membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
Specifications

Specifications

Dystrophin
Polyclonal
Rabbit
Muscle Biology
A synthetic peptide corresponding to C-terminal of human Dystrophin
Immunohistochemistry (Paraffin)
Unconjugated
IVD
Primary
IgG
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit