Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DZIP1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549659

Catalog No. PA549659

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

May participate in spermatogenesis via its interaction with DAZ.
Specifications

Specifications

DZIP1
Polyclonal
PBS with 0.09% sodium azide
Q86YF9
Zinc finger protein DZIP1, DAZ-interacting protein 1/2, DZIP1, DZIP, DZIP2, KIAA0996
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
22873
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
DZIP1
Liquid
DZIP1
KLH conjugated synthetic peptide between 568-596 amino acids from the Central region of human DZIP1.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit