Alfa Aesar™ (E)-1-Pentenylboronic acid pinacol ester, 98%

Manufacturer: Alfa Aesar™ H2774903

Quantity 1g

 View more versions of this product

Catalog No. AAH2774903

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 161395-96-6
    Molecular Formula C11H21BO2
    Formula Weight 196.1
    MDL Number MFCD05663882
    Synonym (E)-2-(1-Pentenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane; 2-Hexyl-4,4,5,5-tetramethyl[1,3,2]dioxaborolane
    Chemical Name or Material (E)-1-Pentenylboronic acid pinacol ester

    Specifications

    Formula Weight 196.1
    Molecular Formula C11H21BO2
    MDL Number MFCD05663882
    CAS 161395-96-6
    Synonym (E)-2-(1-Pentenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane; 2-Hexyl-4,4,5,5-tetramethyl[1,3,2]dioxaborolane
    Health Hazard 2 GHS H Statement
    H315-H319-H335-H227
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Combustible liquid.
    Health Hazard 3 GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Boiling Point 110° to 111°C (28mmHg)
    Quantity 1g
    Flash Point 82°C (180°F)
    Health Hazard 1 Warning
    Chemical Name or Material (E)-1-Pentenylboronic acid pinacol ester
    Assay 98%

    Description

    Applications
    Used as reagents in Suzuki-Miyaura coupling reactions.

    Solubility
    Insoluble in water.

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents. Keep in freezer under -20°C.

    GHS H Statement
    H315-H319-H335-H227
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Combustible liquid.

    GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit