Learn More
3M™ E-A-Rsoft™ Yellow Neons™ One Touch™ Dispenser Refill
Yellow Neons™ Earplugs in a refill container for the One Touch™ Dispenser. Earplugs seal the ear canal with soft, slow-recovery foam for excellent noise reduction and comfort; available in a large size option to comfortably fit in larger ears.
Specifications
Hearing Protection Style Uncorded
Style Roll Down Insertion
Material Polyurethane
Packaging Type Box
Plug Material Foam
View More Specs
Products 2
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-072-324
View Documents
3M™
70071515087
Regular This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
19-072-325
View Documents
3M™
70071521309
Large This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
  • Packaged in convenient Refill Bottles to fit One Touch™ Dispensers (391-1000)
  • Regular-sized earplugs are test compatible with 3M™ E-A-Rfit™ Validation System
  • Available in a large size option to accomodate users with larger ear canals
  • The NRR may overestimate the hearing protection provided during typical use
  • No packaging wasted with each new pair
Specifications
Specifications
Uncorded
Polyurethane
Foam
Tapered
33 dB
Roll Down Insertion
Box
One Touch™ Refill Bottle
Yellow