Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
E-Cadherin Antibody (4A2), PE/Cy7, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP254587PECY7
Description
E-Cadherin Monoclonal specifically detects E-Cadherin in Human samples. It is validated for Flow Cytometry.Specifications
E-Cadherin | |
Monoclonal | |
PE-Cyanine7 | |
PBS with 0.05% Sodium Azide | |
CDH1 | |
Recombinant human E-Cadherin protein (Uniprot: P12830 ) | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 κ |
Flow Cytometry | |
4A2 | |
Flow Cytometry | |
Arc-1, cadherin 1, E-cadherin (epithelial), cadherin 1, type 1, E-cadherin (epithelial), cadherin-1, calcium-dependent adhesion protein, epithelial, CAM 120/80, CD324, CD324 antigen, CDHE, cell-CAM 120/80, ECAD, E-cadherin, Epithelial cadherin, LCAM, UVOE-Cadherin, uvomorulin | |
Mouse | |
Protein A or G purified | |
Cancer, Cell Cycle and Replication, Cellular Markers, Extracellular Matrix, Phospho Specific, Plasma Membrane Markers, Signal Transduction | |
999.0 | |
Store at 4°C in the dark. Do not freeze. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction