Learn More
E-Cadherin Rat anti-Mouse, Alexa Fluor 350, Clone: 114420, R&D Systems™
Rat Monoclonal Antibody
Supplier:  R&D Systems FAB7481U100UG
Catalog No. FAB7481U100
Add to Cart
This item is not returnable. View return policy
Description
Description
E-Cadherin Monoclonal specifically detects E-Cadherin in Mouse samples. It is validated for Flow Cytometry.
Specifications
Specifications
E-Cadherin
Monoclonal
0.2 mg/mL
Flow Cytometry 0.25-1 ug/10^6 cells
P09803
CDH1
Mouse myeloma cell line NS0-derived recombinant mouse E-Cadherin Asp157-Val709 (predicted) Accession # P09803
RUO
999
Mouse
IgG2a
Flow Cytometry
114420
Alexa Fluor 350
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Arc-1, CAD1, cadherin 1, E-cadherin (epithelial), cadherin 1, type 1, E-cadherin (epithelial), cadherin-1, calcium-dependent adhesion protein, epithelial, CAM 120/80, CD324, CD324 antigen, CDH1, CDHE, cell-CAM 120/80, Cell-CAM120/80, ECAD, E-cadherin, ECadherin, Epithelial cadherin, LCAM, L-CAM, UVOE-Cadherin, uvomorulin
Rat
100 μg
Primary
Detects mouse E-Cadherin in direct ELISAs and Western blots.
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)