Learn More
E-Cadherin Rat anti-Mouse, Alexa Fluor 700, Clone: 114420, R&D Systems™
Rat Monoclonal Antibody
Manufacturer:  R&D Systems FAB7481N100UG
Catalog No. FAB7481N100
Add to cart
This item is not returnable. View return policy
Description
Description
E-Cadherin Monoclonal specifically detects E-Cadherin in Mouse samples. It is validated for Flow Cytometry.
Specifications
Specifications
E-Cadherin
Monoclonal
0.2 mg/mL
Flow Cytometry 0.25-1 ug/10^6 cells
P09803
CDH1
Mouse myeloma cell line NS0-derived recombinant mouse E-Cadherin Asp157-Val709 (predicted) Accession # P09803
100 ug
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Monoclonal
Detects mouse E-Cadherin in direct ELISAs and Western blots.
Flow Cytometry
114420
Alexa Fluor 700
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Arc-1, CAD1, cadherin 1, E-cadherin (epithelial), cadherin 1, type 1, E-cadherin (epithelial), cadherin-1, calcium-dependent adhesion protein, epithelial, CAM 120/80, CD324, CD324 antigen, CDH1, CDHE, cell-CAM 120/80, Cell-CAM120/80, ECAD, E-cadherin, ECadherin, Epithelial cadherin, LCAM, L-CAM, UVOE-Cadherin, uvomorulin
Rat
IgG2a
RUO
Primary
999
Mouse
SDS
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)