Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ E. coli Beta Lactamase Recombinant Protein

Highly purified and high bioactivity. Generating reliable and reproducible results. Applications: Functional, SDS-Page, Bioactivity

$131.70 - $356.50

Specifications

Product Type Recombinant Protein
Recombinant Yes
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles
Gene Symbol ampC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB59641100U View Documents Novus Biologicals™
NBP259641100UG
100μg Each for $356.50
Add to cart
 
NB5964120UG View Documents Novus Biologicals™
NBP25964120UG
20μg Each for $131.70
Add to cart
 
Description

Description

A bioactive recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 20-377 of E. coli Beta Lactamase The Recombinant E. coli Beta Lactamase Protein is derived from E. coli. The Recombinant E. coli Beta Lactamase Protein has been validated for the following applications: Functional, SDS-Page, Bioactivity.
Specifications

Specifications

Recombinant Protein
RUO
ampC
20mM Tris-HCl buffer (pH8.0) containing 10% glycerol
E.coli
Unlabeled
Yes
Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles
AmpC, EC 3.4, FLJ14902
E.coli
AC
41.8 kDa

For Research Use Only.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit