Learn More
BioworldSupplier Diversity Partner E.COLI DNAK (HSP70) (RECOM DFS Item
Manufacturer:  BioworldSupplier Diversity Partner 220600571
E.COLI DNAK (HSP70), (RECOMBINANT): Recombinant Dnak Source: E.Coli Description : DnaK, originally identified for its DNA replication by bacteriophage lambda in E. coli is the bacterial hsp70 chaperone. This protein is involved in th...
Catalog No. 50-253-154
Due to product restrictions, please to purchase or view availability for this product.