Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ E. coli eutC Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5117515
Description
This antibody has been tested in direct-ELISA.
The ethanolamine ammonialyase microcompartment of Escherichia coli represents such a class of cytosolic organelles that enables bacteria to survive on small organic molecules such as ethanolamine as the sole source for carbon and nitrogen.
Specifications
E. coli eutC | |
Polyclonal | |
Unconjugated | |
eutC | |
eutC | |
Recombinant Escherichia coli Ethanolamine ammonia-lyase light chain protein (1-295aa). | |
100 μg | |
Primary | |
Bacteria | |
Antibody | |
IgG |
Western Blot | |
3 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
P19636 | |
Rabbit | |
Protein G | |
RUO | |
946925 | |
-20°C or -80°C if preferred | |
Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction