Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ E. coli serotype K99 Pili Monoclonal Antibody (K99 12-349.2), Invitrogen™

Catalog No. p-7245017
Change view
Click to view available options
Quantity:
1 mg
100 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA546082 100 μg
01-671-825 500 μg
PIMA546161 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PIMA546082 Supplier Invitrogen™ Supplier No. MA546082
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA1-10796 detects E. coli K99 pili. These antibodies do not cross-react with k99-negative E.Coli, E.Coli K88ac, E.Coli 987P, Proteus vulgaris, Pseudomonas aeruginosa, Enterbacter aerogenes, Citrobacter freundii, Klebsiella pneumoniae, or strep. agalactiae. MA1-10796 has been successfully used in ELISA applications. The MA1-10796 immunogen is purified E. coli K99 pili.

Escherichia coli (E. coli) are Gram negative enteric bacteria that live in the human gut. Its presence in nature is an indication of human feacal pollution. E. coli also causes human disease such as urinary tract infections and neonatal meningitis. Infection with particular enterotoxigenic strains can lead to life threatening intestinal diseases. E. coli are distinguished immunologically by serotyping based on three types of antigen: the somatic or O antigen which corresponds to terminal sugars on the cell surface lipopolysaccharide, the capsular or K antigens and the flagellar or H antigens.
TRUSTED_SUSTAINABILITY

Specifications

Antigen E. coli serotype K99 Pili
Applications ELISA
Classification Monoclonal
Clone K99 12-349.2
Concentration 1.1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Alias Escherichia coli
Host Species Mouse
Immunogen Purified E. coli K99 pili.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.