Learn More
Invitrogen™ E. coli serotype K99 Pili Monoclonal Antibody (K9912-418.2), Invitrogen™

Description
MA1-10797 detects E. coli K99 pili. MA1-10797 has been successfully used in ELISA applications. The MA1-10797 immunogen is purified E. coli K99 pili.
Specifications
Specifications
| Antigen | E. coli serotype K99 Pili |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | K9912-418.2 |
| Concentration | 0.8 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene Alias | Escherichia coli |
| Host Species | Mouse |
| Immunogen | Purified E. coli K99 pili. |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.