Learn More
E. coli serotype K99 Pili Monoclonal Antibody (K9912-418.2), Invitrogen™
Mouse Monoclonal Antibody
Supplier:  Thermo Scientific MA546163
 View more versions of this product
Catalog No. PIMA546163
Add to Cart
This item is not returnable. View return policy
Description
Description
MA1-10797 detects E. coli K99 pili. MA1-10797 has been successfully used in ELISA applications. The MA1-10797 immunogen is purified E. coli K99 pili.
E. coli serotype K99 Pili Monoclonal antibody specifically detects E. coli serotype K99 Pili in Bacteria samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
E. coli serotype K99 Pili
Monoclonal
∽1 mg/mL
PBS with no preservative; pH 7.4
Mouse
Protein A
RUO
Bacteria
Antibody
IgG1
ELISA
K9912-418.2
Unconjugated
Escherichia coli
Purified E. coli K99 pili.
1 mg
Primary
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Recommendations
Recommendations