Learn More
Invitrogen™ E. coli serotype O Monoclonal Antibody (HEC10-118.1), Invitrogen™

Description
MA1-10696 detects E. coli from E. coli O2a/2b, O7, O11, O18, O44, O75, O112, and O125 samples. MA1-10696 has been successfully used in ELISA applications. The MA1-10696 immunogen is E. coli serotypes: O18, O44, O112, O125.
Specifications
Specifications
| Antigen | E. coli serotype O |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | HEC10-118.1 |
| Concentration | 1.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene Alias | Escherichia coli |
| Host Species | Mouse |
| Immunogen | E. coli serotypes: O18, O44, O112, O125. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.