Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ E. coli serotype O Monoclonal Antibody (HEC10-144.2), Invitrogen™

Catalog No. PIMA546095
Change view
Click to view available options
Quantity:
1 mg
100 μg
500 μg
Catalog No. Quantity
PIMA546095 1 mg
PIMA546016 100 μg
01-671-924 500 μg
3 options
This item is not returnable. View return policy

Catalog No. PIMA546095

Supplier: Invitrogen™ MA546095

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

MA1-10693 detects E. coli from E. coli O2a/2b, O7, O11, O18, O44, O75, O112, O125 samples. MA1-10693 has been successfully used in ELISA applications. The MA1-10693 immunogen is E. coli serotypes: O18, O44, O112, O125.

Escherichia coli (E. coli) are Gram negative enteric bacteria that live in the human gut. Its presence in nature is an indication of human feacal pollution. E. coli also causes human disease such as urinary tract infections and neonatal meningitis. Infection with particular enterotoxigenic strains can lead to life threatening intestinal diseases. E. coli are distinguished immunologically by serotyping based on three types of antigen: the somatic or O antigen which corresponds to terminal sugars on the cell surface lipopolysaccharide, the capsular or K antigens and the flagellar or H antigens.
TRUSTED_SUSTAINABILITY

Specifications

Antigen E. coli serotype O
Applications ELISA
Classification Monoclonal
Clone HEC10-144.2
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.2-7.4
Gene Alias Escherichia coli
Host Species Mouse
Immunogen E. coli serotypes: O18, O44, O112, O125.
Purification Method Protein A
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.