Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
E. coli serotype O Monoclonal Antibody (HEC10-189.4), Invitrogen™

Mouse Monoclonal Antibody
$472.00 - $1160.00
Specifications
| Antigen | E. coli serotype O |
|---|---|
| Clone | HEC10-189.4 |
| Concentration | 0.9 mg/mL |
| Applications | ELISA |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
PIMA546074
|
Invitrogen™
MA546074 |
100 μg |
Each for $472.00
|
|
|||||
01-671-925
![]() |
Invitrogen™
MA110694 |
500 μg |
Each for $632.00
|
|
|||||
|
PIMA546097
|
Invitrogen™
MA546097 |
1 mg |
Each for $1,160.00
|
|
|||||
Description
MA1-10694 detects E. coli from E. coli O2a/2b, O7, O11, O18, O44, O75, O112, and O125 samples. MA1-10694 has been successfully used in ELISA applications. The MA1-10694 immunogen is E. coli serotypes: O18, O44, O112, O125.
Escherichia coli (E. coli) are Gram negative enteric bacteria that live in the human gut. Its presence in nature is an indication of human feacal pollution. E. coli also causes human disease such as urinary tract infections and neonatal meningitis. Infection with particular enterotoxigenic strains can lead to life threatening intestinal diseases. E. coli are distinguished immunologically by serotyping based on three types of antigen: the somatic or O antigen which corresponds to terminal sugars on the cell surface lipopolysaccharide, the capsular or K antigens and the flagellar or H antigens.Specifications
| E. coli serotype O | |
| 0.9 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with no preservative; pH 7.4 | |
| E. coli serotypes: O18, O44, O112, O125. | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles |
| HEC10-189.4 | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Bacteria | |
| Escherichia coli | |
| IgG1 | |
| Protein G | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title