Learn More
Invitrogen™ E-Gel™ 48 Agarose Gels with SYBR™ Safe DNA Gel Stain, 2%
Designed for fast, convenient, and safe DNA sample electrophoresis
$311.00 - $1002.00
Specifications
Compatible Ladders | E-Gel™ 1 Kb Plus Express DNA Ladder |
---|---|
Content And Storage | • Each box contains 8 gels Store strictly at room temperature. Do not freeze. Avoid cold spots near freezers or other uncontrolled environments. |
Gel Percentage | 2% |
Gel Type | E-Gel |
Product Type Specs | Agarose Gel |
Catalog Number | Mfr. No. | Formulation | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Formulation | Quantity | Price | Quantity & Availability | ||||
G820802
![]() |
Invitrogen™
G820802 |
E-Gel: 2% Stain, SYBR Safe | 8 Gels |
Each for $311.00
|
|
G820842
![]() |
Invitrogen™
G820842 |
E-Gel: 2% Stain, SYBR Safe | 4 x 8 Gels |
Pack of 4 for $1,002.00
|
|
Description
Invitrogen E-Gel 48 high-throughput precast agarose gels with SYBR Safe DNA gel stain are designed for fast, convenient, and safe DNA sample electrophoresis. Each E-Gel agarose gel cassette contains all components and stain required for efficient gel separation and analysis—just load your samples and run.
Each gel contains 48 sample wells and 4 marker wells in 1% or 2% high-resolution agarose with a 3.2-cm run length. The various agarose percentages are used to separate DNA fragments of different sizes:
• 1% agarose: DNA fragments between 400 bp and 10 kb
• 2% agarose: DNA fragments between 50 bp and 3 kb
Each cassette is individually labeled with an EAN39 bar code that is recognized by most commercially available robotic bar code readers.
Features of E-Gel 48 gels include:
• High throughput analysis—contains 48 sample lanes and 4 marker lanes per gel
• Automation compatible—suitable for use with multi-channel pipettors and commonly used 8- and 12-pin liquid handling robots
• Scalable—resolve up to 192 sample in one run by combining up to four E-Base Mother and Daughter devices
• Safer choice—contains SYBR Safe DNA stain, a safer alternative to ethidium bromide
• High performance—detect 2–3 ng of sample DNA with a separation time as short as ∼20 minutes
Safer option
Choose SYBR Safe DNA stained gels to minimize exposure to hazardous and mutagenic factors such as ethidium bromide or UV. Precast E-Gel agarose gels with SYBR Safe, in combination with blue-light trans-illumination, offer better sample integrity and a safer work environment.
Unlike ethidium bromide, the DNA stain most commonly incorporated into agarose gels, SYBR Safe DNA Gel Stain is a non-toxic, non-mutagenic DNA stain that is safer for the user, safer for the environment, and can save institutional overhead costs for disposal.
Fast and convenient analysis
E-Gel agarose gels enable fast and high-performance analysis with a detection sensitivity of 2–3 ng of sample DNA per band and a separation time 2–3 fold faster than that of conventional pour-your-own gels.
Each E-Gel agarose gel contains agarose, electrodes, SYBR Safe DNA stain, and a buffer-less ion exchange system packaged inside a dry disposable cassette. E-Gel technology does not require any gel or buffer preparation or gel straining steps. Just load your samples and run.
Nucleic acid markers
For optimal DNA analysis on E-Gel 48 agarose gels we recommend the specially formulated E-Gel 96 High Range DNA Ladder (Cat. No. 12352019) or E-Gel 50 bp DNA Ladder (Cat. No. 10488099). Our high-quality ready-to-use ladders are formulated using individual chromatography-purified DNA fragments and are suitable for storage at room temperature.
Running and imaging devices
High-throughput E-Gel agarose gels require the E-Base Electrophoresis Device for gel running.
Available in a variety of gel formats
E-Gel agarose gels are available in variety of gel percentage, stain, and well formats. The high-throughput E-Gel 96 and 48 agarose gels with SYBR Safe are available in 1% and 2% gel percentages.
Specifications
E-Gel™ 1 Kb Plus Express DNA Ladder | |
2% | |
Agarose Gel | |
48 + 4-well | |
Less hazardous |
• Each box contains 8 gels Store strictly at room temperature. Do not freeze. Avoid cold spots near freezers or other uncontrolled environments. |
|
E-Gel | |
50 bp to 3 kb | |
E-Gel: 2% Stain, SYBR Safe | |
Room Temperature |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.