Learn More
Invitrogen™ E-Gel™ EX Double Comb Agarose Gels, 1%
Invitrogen™ E-Gel™ EX Double Comb precast agarose gels are designed for ultrasensitive and convenient DNA sample electrophoresis in as little as 5–10 minutes.
$278.00 - $1126.00
Specifications
Compatible Ladders | E-Gel™ 1 Kb Plus Express Ladder |
---|---|
Content And Storage | • Each box contains 10 gels Store strictly at room temperature. Do not freeze. Avoid cold spots near freezers or other uncontrolled environments. |
Gel Percentage | 1% |
Gel Type | E-Gel |
Product Type | Agarose Gel |
Description
Invitrogen™ E-Gel™ EX Double Comb precast agarose gels are designed for ultrasensitive and convenient DNA sample electrophoresis in as little as 5–10 minutes. Each E-Gel™ EX Double Comb gel cassette contains all components and stain required for efficient gel separation and analysis—just load your samples and run. E-Gel™ EX Double Comb gels are ideal for analyzing PCR products, restriction digests, plasmid preparations, or genotyping samples.
Features of E-Gel™ EX Double Comb Agarose Gels include:
- Instantaneous setup—precast E-Gel™ agarose gels are ready for use when you are, and do not require gel preparation or additional buffers
- High sensitivity—detect 1 ng of sample DNA
- Rapid analysis—complete DNA sample separation in 5–10 minutes
- In-process control—view DNA sample migration in real time
- Easy sample access—openable cassette design allows easy in-gel access for convenient DNA sample excision or transfer to membranes
- For DNA—ideal for DNA sample analysis
Ultra-sensitive detection of DNA
E-Gel™ EX Double Comb gels include SYBR Gold DNA stain and were developed for ultimate sensitivity, demonstrating up to five-fold greater sensitivity than comparable gels containing ethidium bromide. This superior sensitivity allows you to use lower amounts of sample, saving time and money.
Fast and convenient analysis
E-Gel™ EX Double Comb gels are our fastest-resolving agarose gels, enabling sample analysis in as little as 5-10 minutes. Each E-Gel™ EX Double Comb gel contains agarose, electrodes, SYBR Gold DNA stain, and a bufferless ion exchange system packaged inside a dry disposable cassette. E-Gel™ technology does not required any gel or buffer preparation or gel staining steps. Just load your samples and run.
Live monitoring of DNA migration
E-Gel™ EX Double Comb gels include SYBR Gold II DNA gel stain with an excitation wavelength in the blue-light spectrum. When used on the E-Gel™ Power Snap Electrophoresis Device with integrated blue-light trans-illuminator, less-hazardous real-time monitoring of DNA migration is enabled without risk to eyes or possible damage of the sample, unlike UV illumination.
Easy access to the gel
E-Gel™ EX Double Comb cassettes are designed for convenient opening with a Gel Knife (Cat. No. EI9010), for easy excision of specific bands or transfer of the gel to a membrane for southern blot analysis.
Nucleic acid markers
For optimal DNA analysis on E-Gel™ EX Double Comb gels, we recommend specially formulated E-Gel™ 1 Kb Plus Express DNA Ladder (Cat. No. 10488091), E-Gel™ 50 bp DNA Ladder (Cat. No. 10488099), or E-Gel™ 96 High Range DNA Marker (Cat. No. 12352019). Our high-quality ready-to-use ladders are formulated using individual chromatography-purified DNA fragments and are suitable for storage at room temperature.
Running and imaging devices
E-Gel™ EX Double Comb gels require the Invitrogen E-Gel™ Power Snap Electrophoresis System for gel running and viewing. The system consists of an electrophoresis device and a camera for fast and convenient E-Gel™ EX Double Comb gel separation and analysis.
Available in a variety of gel formats
E-Gel™ agarose gels are available in variety of gel percentage, stain, and well formats. The 22-well E-Gel™ EX Double Comb gels are available in 1% and 2% gel
Specifications
E-Gel™ 1 Kb Plus Express Ladder | |
1% | |
Agarose Gel | |
SYBR™ Gold | |
Double Comb |
• Each box contains 10 gels Store strictly at room temperature. Do not freeze. Avoid cold spots near freezers or other uncontrolled environments. |
|
E-Gel | |
100 to 5000 bp | |
22-well |
For Research Use Only. Not for use in diagnostic procedures.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.