Learn More
SiliCycle™ E-PAK™ Cartridges, Commercial Scale, SiliaCarb™ C-CA Activated Carbon
E-PAK™ Cartridges, Commercial Scale, SiliaCarb™ C-CA Activated Carbon is a chemically activated wood based powdered activated carbon conveniently packaged in a 16.5 X 100 cm cartridge.
Supplier:  SiliCycle™ CSCCA1650
 View more versions of this product
Catalog No. 01-182-016
Add to cart
Description
Description
  • Testing with loose media can be done with samples as small as a few milliliters and are normally done in advance of cartridge testing to identify the formula with the highest capacity to remove contaminants with the least loss of product.
  • Testing has shown good correlation between the performance of lab, pilot and commercial size cartridges.
  • Activated Carbon weight: 3.4 kg (16.5 x 50 cm), 6.5 kg (16.5 x 100 cm)
Specifications
Specifications
Commercial Scale E-PAK Cartridge
Black
1 Pk.
Chemically Activated Carbon
Removal of precious metal catalysts, pigments and reaction contaminants
50 x 16.5 cm
6 to 8