Learn More
SiliCycle™ E-PAK™ Cartridges, Commercial Scale, SiliaCarb™ C-VW Activated Carbon
E-PAK™ Cartridges, Commercial Scale, SiliaCarb™ C-VW Activated Carbon is a virgin wood based catalytic activated carbon conveniently packaged in a cartridge.
Supplier: SiliCycle™ CSCVW1650
Description
- Testing with loose media can be done with samples as small as a few milliliters and are normally done in advance of cartridge testing to identify the formula with the highest capacity to remove contaminants with the least loss of product.
- Testing has shown good correlation between the performance of lab, pilot and commercial size cartridges.
- Activated Carbon weight: 3.4 kg (16.5 x 50 cm), 6.5 kg (16.5 x 100 cm)
Specifications
Commercial Scale E-PAK Cartridge | |
Black | |
1 Pk. | |
Virgin Wood Carbon |
Removal of precious metal catalysts, pigments and reaction contaminants | |
50 x 16.5 cm | |
6 to 8 |