Learn More
SiliCycle™ E-PAK™ Cartridges, Commercial Scale, SiliaFlash™ 60 - 200 μm, 60 Å
E-PAK™ Cartridges, Commercial Scale, SiliaFlash™ 60 - 200 μm, 60 Å irregular silica gels, F60, are conveniently packaged in a cartridge.
Supplier: SiliCycle™ CSR10040B1650
Description
- Narrow particle size distribution
- Superior resolution
- Suitable for use with complex matrices
- Suitable for organic molecules (MW <1000 Dalton)
- Testing has shown a good correlation between the performance of lab, pilot, and commercial size cartridges.
Specifications
E-PAK Cartridge | |
50 x 16.5 cm | |
Irregular |